Learn More
Tetraethylthiuram disulfide, 97%, Thermo Scientific Chemicals
£17.40 - £298.00
Chemical Identifiers
CAS | 97-77-8 |
---|---|
Molecular Formula | C10H20N2S4 |
Molecular Weight (g/mol) | 296.524 |
MDL Number | MFCD00009048 |
InChI Key | AUZONCFQVSMFAP-UHFFFAOYSA-N |
Synonym | disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd |
PubChem CID | 3117 |
ChEBI | CHEBI:4659 |
IUPAC Name | diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate |
SMILES | CCN(CC)C(=S)SSC(=S)N(CC)CC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11403950
|
Thermo Scientific Alfa Aesar
B20721.22 |
100 g |
£17.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11475576
|
Thermo Scientific Alfa Aesar
B20721.30 |
250 g |
£28.70
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11465576
|
Thermo Scientific Alfa Aesar
B20721.0B |
1000 g |
£86.60
1000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11485576
|
Thermo Scientific Alfa Aesar
B20721.0I |
5000 g |
£298.00
5000g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
Dopamine beta-hydroxylase inhibitor
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemische Identifikatoren
97-77-8 | |
296.524 | |
AUZONCFQVSMFAP-UHFFFAOYSA-N | |
3117 | |
diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate |
C10H20N2S4 | |
MFCD00009048 | |
disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd | |
CHEBI:4659 | |
CCN(CC)C(=S)SSC(=S)N(CC)CC |
Spezifikation
97-77-8 | |
117°C (17 mmHg) | |
MFCD00009048 | |
1712560 | |
disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd | |
CCN(CC)C(=S)SSC(=S)N(CC)CC | |
296.524 | |
CHEBI:4659 | |
97% |
69°C to 72°C | |
C10H20N2S4 | |
UN3077 | |
14,3364 | |
AUZONCFQVSMFAP-UHFFFAOYSA-N | |
diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate | |
3117 | |
296.54 | |
Tetraethylthiuram disulfide |
Sicherheit und Handhabung
GHS H Statement
H373-H302-H317
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
May cause an allergic skin reaction.
GHS P Statement
P260-P261-P280-P301+P312-P363-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wash contaminated clothing before reuse.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
missing translation for 'rtecsNumber' : JO1225000
missing translation for 'tsca' : TSCA
RUO – Research Use Only