missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylthiuram disulfide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Melting Point | 70.0°C to 74.0°C |
Color | Yellow |
Boiling Point | 117°C (23.0 hPa) |
Loss on Drying | 0.3% max. (60°C) |
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (HPLC) |
Packaging | Plastic bottle |
Linear Formula | [(C2H5)2NCS2-]2 |
Show More |
Safety and Handling
Product Identifier
- Tetraethylthiuram disulfide
- Warning
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Respiratory sensitiser Category 1
- Specific target organ toxicity after repeated exposure Category 2
- H317-May cause an allergic skin reaction.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'