missing translation for 'onlineSavingsMsg'
Learn More
Tetrafluoroboric acid, 48% min w/w aq. soln., Thermo Scientific Chemicals Product Code.: 11327147

Tetrafluoroboric acid, 48% min w/w aq. soln., Thermo Scientific Chemicals

Product Code. 11327147
50 g
Click to view available options
Quantity:
50 g
500 g
4 x 500 g
This item is not returnable. View return policy

Product Code. 11327147

Brand: Thermo Scientific Alfa Aesar 011484.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tetrafluoroboric acid is used in conjunction with various methods of cleaning and treat the surface of substrates. It is used as radiotracers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16872-11-0
MDL Number MFCD00011345
Synonym fluoroboric acid, fluoboric acid, tetrafluoroboric acid, hydrogen tetrafluoroborate, borofluoric acid, hydrofluoboric acid, borate 1-, tetrafluoro-, hydrogen, hydron tetrafluoroborate, hydrogen tetrafluoroborate 1-, unii-h429wz9fbqShow MoreShow Less

Specifications

Boiling Point 130°C (decomposition)
Density 1.4
Quantity 50 g
Physical Form Liquid
Solubility Information Miscible with water.
Formula Weight 87.81
Chemical Name or Material Tetrafluoroboric Acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP07L037 Certificate TypeCertificate of Analysis Date10/03/2025 Catalog Number011484.36,011484.57,011484.18
Lot NumberM13L013 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number011484.36,011484.57,011484.18
Lot NumberY12K032 Certificate TypeCertificate of Analysis Date26/12/2024 Catalog Number011484.36,011484.57,011484.18
Lot NumberU14K027 Certificate TypeCertificate of Analysis Date29/08/2024 Catalog Number011484.36,011484.57,011484.18
Lot NumberR03K037 Certificate TypeCertificate of Analysis Date08/05/2024 Catalog Number011484.36,011484.57,011484.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrafluoroboric acid, 48% min w/w aq. soln., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.