missing translation for 'onlineSavingsMsg'
Learn More
Tetrafluoroboric acid, ca 50% w/w aq. soln., Thermo Scientific Chemicals Product Code.: 11368915

Tetrafluoroboric acid, ca 50% w/w aq. soln., Thermo Scientific Chemicals

Product Code. 11368915
100 g
Click to view available options
Quantity:
100 g
500 g
2500 g
This item is not returnable. View return policy

Product Code. 11368915

Brand: Thermo Scientific Alfa Aesar L14037.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tetrafluoroboric acid is used in conjunction with various methods of cleaning and to treat the surface of substrates. It is also used as a radiotracer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetrafluoroboric acid is used in conjunction with various methods of cleaning and to treat the surface of substrates. It is also used as a radiotracer.

Solubility
Miscible with water.

Notes
Incompatible with strong bases, oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16872-11-0
MDL Number MFCD00011345
Synonym fluoroboric acid, fluoboric acid, tetrafluoroboric acid, hydrogen tetrafluoroborate, borofluoric acid, hydrofluoboric acid, borate 1-, tetrafluoro-, hydrogen, hydron tetrafluoroborate, hydrogen tetrafluoroborate 1-, unii-h429wz9fbqShow MoreShow Less

Specifications

Boiling Point 130°C (decomposition)
Concentration ≈50% w/w aq. soln.
Density 1.4
Odor Pungent
Quantity 100 g
Solubility Information Miscible with water.
Formula Weight 87.81
Concentration or Composition (by Analyte or Components) ≈50% w/w aq. soln.
Chemical Name or Material Tetrafluoroboric acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberN05L010 Certificate TypeCertificate of Analysis Date07/02/2025 Catalog NumberL14037.0E,L14037.36,L14037.22
Lot NumberT03K002 Certificate TypeCertificate of Analysis Date19/07/2024 Catalog NumberL14037.0E,L14037.36,L14037.22
Lot Number10234856 Certificate TypeCertificate of Analysis Date18/04/2024 Catalog NumberL14037.0E,L14037.36,L14037.22
Lot NumberS02J049 Certificate TypeCertificate of Analysis Date25/10/2023 Catalog NumberL14037.0E,L14037.36,L14037.22
Lot NumberM25J020 Certificate TypeCertificate of Analysis Date01/02/2023 Catalog NumberL14037.0E,L14037.36,L14037.22
compliance-icons compliance-icons
Product Identifier
  • Tetrafluoroboric acid, ca 50% w/w aqueous solution
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : fluoroboric acid, boric acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrafluoroboric acid, ca 50% w/w aq. soln., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.