missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrafluoroboric Acid Solution, Honeywell Fluka™
Click to view available options
Quantity:
25 g
500 g
6 x 500 g
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Concentration | 48wt. % in H2O |
Density | 1.4g/mL at 25°C |
Vapor Pressure | 5mmHg (20°C) |
Quantity | 25 g |
Linear Formula | HBF4 |
UN Number | 1775 |
Formula Weight | 87.81g/mol |
Chemical Name or Material | Tetrafluoroboric Acid Solution |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'