missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydro-2H-pyran-4-carboxylic acid, 97+%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
Specifications
Assay Percent Range | 97% |
Quantity | 1 g |
Formula Weight | 130.14 |
Percent Purity | 97% |
Chemical Name or Material | Tetrahydro-2H-pyran-4-carboxylic acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tetrahydro-2H-pyran-4-carboxylic acid, 97+%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction