missing translation for 'onlineSavingsMsg'
Learn More
Tetrahydrofuran-3-sulfonyl chloride, Thermo Scientific™ Product Code.: 10311825

Tetrahydrofuran-3-sulfonyl chloride, Thermo Scientific™

Product Code. 10311825
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10311825

Brand: Thermo Scientific Maybridge CC76003DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1207346-29-9
Molecular Formula C4H7ClO3S
Molecular Weight (g/mol) 170.607
InChI Key MVZIUOJNVVIOEY-UHFFFAOYSA-N
Synonym tetrahydrofuran-3-sulfonyl chloride, chlorooxolan-3-ylsulfone
PubChem CID 49762850
IUPAC Name oxolane-3-sulfonyl chloride
SMILES C1COCC1S(=O)(=O)Cl

Specifications

Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 170.61
Percent Purity 97%
Chemical Name or Material Tetrahydrofuran-3-sulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrahydrofuran-3-sulfonyl chloride, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.