Learn More
Tetrahydrofuran, 99.5%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™, Thermo Scientific Chemicals
£24.00 - £147.00
Chemical Identifiers
CAS | 109-99-9, 128-37-0 |
---|---|
Molecular Formula | C4H8O |
Molecular Weight (g/mol) | 72.11 |
MDL Number | MFCD00005356 |
InChI Key | WYURNTSHIVDZCO-UHFFFAOYSA-N |
Synonym | tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano |
PubChem CID | 8028 |
ChEBI | CHEBI:26911 |
IUPAC Name | oxolane |
SMILES | C1CCOC1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10292182
|
Thermo Scientific Acros
348451000 |
100 mL |
£24.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10641304
|
Thermo Scientific Acros
348455000 |
500 mL |
£49.80
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10798552
|
Thermo Scientific Acros
348450010 |
1 L |
£85.20
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10256061
|
Thermo Scientific Acros
348450025 |
2.5 L |
£147.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
109-99-9, 128-37-0 | |
72.11 | |
WYURNTSHIVDZCO-UHFFFAOYSA-N | |
8028 | |
oxolane |
C4H8O | |
MFCD00005356 | |
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
CHEBI:26911 | |
C1CCOC1 |
Specifications
-108.4°C | |
Colorless | |
66.0°C | |
Liquid | |
99.5% | |
C4H8O | |
0.55 mPa.s (20°C) | |
0.005% Max. (Coulometric) | |
15, 9356 | |
Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone | |
C1CCOC1 | |
72.11 | |
CHEBI:26911 | |
99.5% | |
Tetrahydrofuran, Water <50 ppm, stabilized |
109-99-9, 128-37-0 | |
0.8800g/mL | |
−21°C | |
1.407 | |
0.88 | |
MFCD00005356 | |
17, 10 | |
01,1140; 02,398; 03,278; 05,649; 06,570 | |
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano | |
WYURNTSHIVDZCO-UHFFFAOYSA-N | |
oxolane | |
8028 | |
72.11 | |
Extra Dry over Molecular Sieve |
Safety and Handling
GHS P Statement May cause respiratory irritation. Causes serious eye irritation. Highly flammable liquid and vapor. May form explosive peroxides. Suspected of causing cancer.
GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water. Keep container tightly closed.
Danger
- Tetrahydrofuran
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only