missing translation for 'onlineSavingsMsg'
Learn More
Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™, package of 4x25ML bottles Product Code.: 12317083

Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™, package of 4x25ML bottles

Product Code. 12317083
100 mL
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy

Product Code. 12317083

Brand: Thermo Scientific Acros 448361000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-99-9 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -108.4°C
CAS 109-99-9, 128-37-0
Color Tan to White
Density 0.8800g/mL
Boiling Point 66.0°C
Quantity 100 mL
Flash Point −21°C
Identification Pass Test
Physical Form Solid
Refractive Index 1.407
Assay Percent Range 99.85%
Residue after Evaporation 0.001 % max.
Specific Gravity 0.88
Molecular Formula C4H8O
MDL Number MFCD00005356
Viscosity 0.55 mPa.s (20°C)
Water 0.005% Max. (Coulometric)
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
Solubility Information Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
SMILES C1CCOC1
IUPAC Name oxolane
Molecular Weight (g/mol) 72.11
PubChem CID 8028
ChEBI CHEBI:26911
Formula Weight 72.11
Percent Purity 99.85%
Grade Extra Dry
Chemical Name or Material Tetrahydrofuran, Stabilized
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2499266 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number448361000
Lot Number2403119 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number448361000
Lot Number2413357 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number448361000
Lot Number2408067 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number448361000
Lot Number2417640 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number448361000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetrahydrofuran
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrahydrofuran, 99.85%, Extra Dry, stabilized, AcroSeal™, package of 4x25ML bottles >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.