missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran, 99%, stab. with 250-350ppm BHT, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 mL
1000 mL
2500 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetrahydrofuran is used as a solvent in Grignard reagent formation processes, pharmaceutical steroids and in the preparation of organometallic reagents. It is used for HPLC, spectrophotometry and environmental testing. It is used as a resin solvent in flexographic inks for plastics coating deposition for audio and video tapes. It is used in PVC and CPVC pipe cements, polyurethane coatings and in PVC film castings.
Solubility
Miscible with water.
Notes
Highly flammable. Oxidizes readily in air to form unstable peroxides. THF exhibits high volatility and an extremely low freezing point. Keep the container tightly closed and sealed until ready for use.
Tetrahydrofuran is used as a solvent in Grignard reagent formation processes, pharmaceutical steroids and in the preparation of organometallic reagents. It is used for HPLC, spectrophotometry and environmental testing. It is used as a resin solvent in flexographic inks for plastics coating deposition for audio and video tapes. It is used in PVC and CPVC pipe cements, polyurethane coatings and in PVC film castings.
Solubility
Miscible with water.
Notes
Highly flammable. Oxidizes readily in air to form unstable peroxides. THF exhibits high volatility and an extremely low freezing point. Keep the container tightly closed and sealed until ready for use.

Specifications
Specifications
Melting Point | -108°C |
Density | 0.889 |
Boiling Point | 66°C |
Quantity | 1000 mL |
Flash Point | −17°C (1°F) |
Odor | Ether-like |
Refractive Index | 1.407 |
UN Number | UN2056 |
Beilstein | 102391 |
Merck Index | 14,9211 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10253646 | Certificate of Analysis | 10/01/2025 | L13304.AU,L13304.0F,L13304.AP |
2411901 | Certificate of Analysis | 26/11/2024 | L13304.AU,L13304.0F,L13304.AP |
2411902 | Certificate of Analysis | 17/10/2024 | L13304.AU,L13304.0F,L13304.AP |
2405221 | Certificate of Analysis | 30/08/2024 | L13304.AU,L13304.0F,L13304.AP |
2486055 | Certificate of Analysis | 30/08/2024 | L13304.AU,L13304.0F,L13304.AP |
Safety and Handling



Product Identifier
- Tetrahydrofuran
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Tetrahydrofuran
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tetrahydrofuran, 99%, stab. with 250-350ppm BHT, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction