missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran, ACS Reagent, contains 250 ppm BHT as inhibitor, ≥99.0%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Melting Point | -108°C (literature) |
CAS Max % | <100.0000% |
Linear Formula | C4H8O |
UN Number | UN2056 |
Beilstein | 102391 |
Formula Weight | 72.11 |
Percent Purity | >99% |
Grade | ACS Reagent |
Density | 0.889 g/mL at 25 °C(lit.) |
Boiling Point | 65°C |
Show More |
Safety and Handling
Product Identifier
- Tetrahydrofuran
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H351-Suspected of causing cancer.
- EUH019-May form explosive peroxides.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'