missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran Anhydrous unstabilised, Honeywell™ Riedel de-Haen

Catalog Number 15807371
Click to view available options
Quantity:
100 mL
2.5 L
This item is not returnable. View return policy
109-99-9
C4H8O
72.11
MFCD00005356
WYURNTSHIVDZCO-UHFFFAOYSA-N
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
8028
CHEBI:26911
oxolane
C1CCOC1
This item is not returnable. View return policy
109-99-9
C4H8O
72.11
MFCD00005356
WYURNTSHIVDZCO-UHFFFAOYSA-N
tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
8028
CHEBI:26911
oxolane
C1CCOC1

Contain less that 50 ppm water. Honeywell™ Riedel de-Haen Anhydrous Solvents are packaged in specially designed septum-sealed bottles to maintain low water content and ensure product integrity.

  • These solvents are specially produced to a water specification of less than 50 ppm (<0.0050%).
  • Many solvents go well beyond that and are routinely produced to lower than 5 ppm (<0.0005%).
  • All packaging steps and materials are especially designed to retain solvent dryness and ensure product integrity. Before filling, amber glass bottles are oven-dried to remove all traces of water, then filled under dry nitrogen and sealed with a special cap.
  • The water content itself is strictly controlled by coulometric Karl Fischer analysis (Hydranal™) and actual results are printed on the label.
  • This line is packaged in specially designed septum-sealed bottles to maintain low water content and ensure product integrity
TRUSTED_SUSTAINABILITY
UN Number 2056
Formula Weight 72.11
Percent Purity >99.9%
Density 0.89 g/cm3 (20°C)
Quantity 2.5 L
Flash Point −21°C
Packaging Septum-sealed Bottle
Chemical Name or Material Tetrahydrofuran
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'