missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, BAKER ANALYZED, J.T. Baker™

Product Code. 10455381 Shop All J.T.Baker Products
Click to view available options
Quantity:
1 L
5 L
25 L
Packaging:
EcoTainer Metal Safety Can
Glass
Steel
Unit Size:
1 L
25 L
5 L
This item is not returnable. View return policy

Product Code. 10455381

Brand: JT Baker 8075.5000

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

High quality grade chemical stabilized with 0.025% BHT for laboratory and specialized industrial use.

  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Melting Point -108.5°C
Color Colorless
Density 0.880 to 0.883 g/mL at 25°C
Boiling Point 65°C to 67°C
Quantity 5 L
Explosion Limit 1.5 - 12 Vol %
Flash Point −21°C
Ignition Point 230°C
Packaging EcoTainer Metal Safety Can
Physical Form Liquid
Residue after Evaporation Max. 0.002%
Linear Formula OCH2CH2CH2CH2
Water Max. 0.02%
Solubility Information Fully miscible in water
Formula Weight 72.11g/mol
Percent Purity ≥99%
Grade BAKER ANALYZED
Chemical Name or Material Tetrahydrofuran
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.