missing translation for 'onlineSavingsMsg'
Learn More
Tetrahydrofuran-D8, for NMR, 99.5 atom % D, Thermo Scientific Chemicals Product Code.: 10712801

Tetrahydrofuran-D8, for NMR, 99.5 atom % D, Thermo Scientific Chemicals

Product Code. 10712801
10 mL, Glass Bottle
Click to view available options
Quantity:
1 mL
5 mL
10 mL
25 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10712801

Brand: Thermo Scientific Acros 180130100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1693-74-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 80.16
MDL Number MFCD00044238
InChI Key WYURNTSHIVDZCO-SVYQBANQSA-N
Synonym tetrahydrofuran-d8, deuterated thf, octadeuterotetrahydrofuran, deuterated tetrahydrofuran, tetrahydrofuran-d8 thf-d8, tetrahydrofuran-d8, ≥99.5 atom % d, 2h4 tetrahydro 2h4 furan, thf-d8, 2h8 tetrahydrofuran, furan-d4-, tetrahydro-d4Show MoreShow Less
PubChem CID 80290
SMILES [2H]C1([2H])OC([2H])([2H])C([2H])([2H])C1([2H])[2H]

Specifications

Physical Form Liquid
Color Colorless
Density 0.9800g/mL
Deuterium (D) 99.4% min.
Packaging Glass Bottle
Water 0.06% max. (Coulometric)
Quantity 10 mL
IUPAC Name 2,2,3,3,4,4,5,5-octadeuteriooxolane
Formula Weight 80.17
Grade NMR
Chemical Name or Material Tetrahydrofuran-D8, 99.5 atom % D
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468897 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number180130250,180130010,180130100,180130050
Lot NumberA0467322 Certificate TypeCertificate of Analysis Date05/11/2024 Catalog Number180130250,180130010,180130100,180130050
Lot NumberA0465715 Certificate TypeCertificate of Analysis Date25/10/2024 Catalog Number180130250,180130010,180130100,180130050
Lot NumberA0466290 Certificate TypeCertificate of Analysis Date23/10/2024 Catalog Number180130250,180130010,180130100,180130050
Lot NumberA0465716 Certificate TypeCertificate of Analysis Date16/10/2024 Catalog Number180130250,180130010,180130100,180130050
compliance-icons compliance-icons
Product Identifier
  • Tetrahydrofuran-D8
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrahydrofuran-D8, for NMR, 99.5 atom % D, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.