Learn More
Tetrahydrofuran, Fluka™
Analytical standard, reference material
Brand: Honeywell Fluka R1588-5ML
Additional Details : CAS Number : 128-37-0 Weight : 0.00440kg
Description
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Specifications
Tetrahydrofuran | |
109-99-9 | |
Colorless | |
Analytical Standard | |
-108°C |
Glass Bottle | |
5 mL | |
72.11 | |
Neutral | |
65°C (1.013 hPa) |