missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran, Puriss. p.a., Honeywell Burdick & Jackson™
Click to view available options
177 kg
Specifications
Melting Point | -108°C |
CAS | 109-99-9 |
Color | Colorless |
Molecular Formula | C4H8O |
MDL Number | MFCD00005356 |
Beilstein | 102391 |
Synonym | THF, Oxolane, 1,4-Epoxybutane, Butylene oxide, Diethylene oxide, Tetra-methylene oxide |
InChI Key | WYURNTSHIVDZCO-UHFFFAOYSA-N |
SMILES | C1CCOC1 |
IUPAC Name | oxolane |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'