missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, ReagentPlus™, ≥99.0%, Honeywell™

Product Code. 15611590 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
10 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
Unit Size:
1 L
10 L
2.5 L
20 L
This item is not returnable. View return policy

Product Code. 15611590

Brand: Honeywell Research Chemicals 17881010L

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Reagent Grade, contains 250 ppm BHT as inhibitor,≥99.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Melting Point -108°C
Color Colorless
Density 0.889g/mL at 25°C
Boiling Point 65°C
Quantity 10 L
Explosion Limit 0.118
Flash Point −17°C (1.4°F)
Ignition Point 610°F
Packaging Stainless Steel Can
Physical Form Liquid
Refractive Index n20/D 1.407(lit.)
Vapor Pressure 114mmHg (15°C), 143mmHg (20°C)
UN Number UN2056
Beilstein 102391
Formula Weight 72.11g/mol
Percent Purity ≥99.0%
Grade ReagentPlus
Chemical Name or Material Tetrahydrofuran, Contains 250 ppm BHT as inhibitor
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.