missing translation for 'onlineSavingsMsg'
Learn More
Tetrahydrofuran stabilised, BAKER ANALYZED™, J.T. Baker™ Product Code.: 10027480
compliance-icons compliance-icons

Tetrahydrofuran stabilised, BAKER ANALYZED™, J.T. Baker™

Product Code. 10027480
2.5 L
Click to view available options
2.5 L
This item is not returnable. View return policy

Product Code. 10027480

Brand: JT Baker 8075.2500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Laboratory Reagent

Specifications

Melting Point -108.5°C
CAS 109-99-9
Color Passes Test
Density 0.888 g/cm³ (20°C)
Boiling Point 66&tdeg;C (1013 hPa)
Quantity 2.5 L
Flash Point -21.5°C
Assay Percent Range min. 99%
Residue after Evaporation max. 0.002%
Molecular Formula C4H8O
MDL Number MFCD00005356
Water max. 0.02%
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
SMILES C1CCOC1
IUPAC Name oxolane
Molecular Weight (g/mol) 72.11
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrahydrofuran stabilised, BAKER ANALYZED™, J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.