missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofurfuryl acrylate, tech. 90%, stab., Thermo Scientific Chemicals

Product Code. 11318365
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2399-48-6
Molecular Formula C8H12O3
Molecular Weight (g/mol) 156.181
MDL Number MFCD00014516
InChI Key YNXCGLKMOXLBOD-UHFFFAOYSA-N
Synonym tetrahydrofurfuryl acrylate, acrylic acid, tetrahydrofurfuryl ester, 2-propenoic acid, tetrahydro-2-furanyl methyl ester, sartomer 285, sartomer 302, viscoat 150, kayarad tc 101, light acrylate thf-a, thf-a, 2-propenoic acid, tetrahydrofurfuryl ester
PubChem CID 94232
IUPAC Name oxolan-2-ylmethyl prop-2-enoate
SMILES C=CC(=O)OCC1CCCO1

Specifications

Density 1.03
Boiling Point 88°C to 89°C (9 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.455
Quantity 250 g
UN Number UN3334
Beilstein 113115
Formula Weight 156.18
Percent Purity 90%
Grade Technical
Chemical Name or Material Tetrahydrofurfuryl acrylate, stabilized with 500ppm hydroquinone
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'