Learn More
Tetrahydropyran-4-carboxylic acid, 98%, Thermo Scientific Chemicals
£37.40 - £142.00
Chemical Identifiers
CAS | 5337-03-1 |
---|---|
Molecular Formula | C6H10O3 |
Molecular Weight (g/mol) | 130.14 |
MDL Number | MFCD00031016 |
InChI Key | AVPKHOTUOHDTLW-UHFFFAOYSA-N |
Synonym | tetrahydro-2h-pyran-4-carboxylic acid, tetrahydropyran-4-yl-carboxylic acid, tetrahydropyran-4-carboxylic acid, tetrahydro-pyran-4-carboxylic acid, 2h-pyran-4-carboxylic acid, tetrahydro, 4-carboxytetrahydro-2h-pyran, 3,4,5,6-tetrahydro-2h-pyran-4-carboxylic acid, 2h-3,4,5,6-tetrahydropyran-4-carboxylic acid, pubchem9730, acmc-2097ha |
PubChem CID | 219302 |
IUPAC Name | oxane-4-carboxylic acid |
SMILES | C1COCCC1C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15411509
|
Thermo Scientific Alfa Aesar
H64340.06 |
5 g |
£37.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15421509
|
Thermo Scientific Alfa Aesar
H64340.14 |
25 g |
£142.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5337-03-1 | |
130.14 | |
AVPKHOTUOHDTLW-UHFFFAOYSA-N | |
219302 | |
C1COCCC1C(=O)O |
C6H10O3 | |
MFCD00031016 | |
tetrahydro-2h-pyran-4-carboxylic acid, tetrahydropyran-4-yl-carboxylic acid, tetrahydropyran-4-carboxylic acid, tetrahydro-pyran-4-carboxylic acid, 2h-pyran-4-carboxylic acid, tetrahydro, 4-carboxytetrahydro-2h-pyran, 3,4,5,6-tetrahydro-2h-pyran-4-carboxylic acid, 2h-3,4,5,6-tetrahydropyran-4-carboxylic acid, pubchem9730, acmc-2097ha | |
oxane-4-carboxylic acid |
Specifications
5337-03-1 | |
White | |
MFCD00031016 | |
tetrahydro-2h-pyran-4-carboxylic acid, tetrahydropyran-4-yl-carboxylic acid, tetrahydropyran-4-carboxylic acid, tetrahydro-pyran-4-carboxylic acid, 2h-pyran-4-carboxylic acid, tetrahydro, 4-carboxytetrahydro-2h-pyran, 3,4,5,6-tetrahydro-2h-pyran-4-carboxylic acid, 2h-3,4,5,6-tetrahydropyran-4-carboxylic acid, pubchem9730, acmc-2097ha | |
C1COCCC1C(=O)O | |
130.14 | |
130.14 | |
Solid |
87°C to 89°C | |
C6H10O3 | |
112249 | |
AVPKHOTUOHDTLW-UHFFFAOYSA-N | |
oxane-4-carboxylic acid | |
219302 | |
98% | |
Tetrahydropyran-4-carboxylic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only