missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydropyran, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Description
It is is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuff. They are also used as important solvents, as chemical intermediate and as monomer for ring-opening polymerization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuff. They are also used as important solvents, as chemical intermediate and as monomer for ring-opening polymerization.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Oxidizing agents, Strong acids. May form peroxides in the absence of inhibitors. Light sensitive
It is is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and dyestuff. They are also used as important solvents, as chemical intermediate and as monomer for ring-opening polymerization.
Solubility
It is soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Oxidizing agents, Strong acids. May form peroxides in the absence of inhibitors. Light sensitive
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -49°C |
Density | 0.882 |
Boiling Point | 87°C to 89°C |
Flash Point | −15°C (5°F) |
Refractive Index | 1.421 |
Quantity | 100 mL |
UN Number | UN3271 |
Beilstein | 102436 |
Merck Index | 14,9217 |
Solubility Information | It is soluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'