missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrothiopyran-4-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10665974
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10665974

Brand: Thermo Scientific Maybridge CC61901DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 89489-53-2
Molecular Formula C6H10O2S
Molecular Weight (g/mol) 146.20
MDL Number MFCD09879921
InChI Key SCKSQOIXSGPVJS-UHFFFAOYSA-N
Synonym tetrahydro-2h-thiopyran-4-carboxylic acid, tetrahydrothiopyran-4-carboxylic acid, 2h-thiopyran-4-carboxylic acid, tetrahydro, 2h-thiopyran-4-carboxylicacid, tetrahydro, 2h-thiopyran-4-carboxylicacid,tetrahydro-9ci, acmc-209vjr, tetrahydro-thiopyran-4-carboxylic acid, 2h-thiopyran-4-carboxylic acid, tetrahydro-9ci
PubChem CID 21598642
IUPAC Name thiane-4-carboxylic acid
SMILES OC(=O)C1CCSCC1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 146.21
Percent Purity 97%
Chemical Name or Material Tetrahydrothiopyran-4-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.