missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrothiopyran-4-ylmethanol, 97%, Thermo Scientific™

Product Code. 10693075
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 100277-27-8
Molecular Formula C6H12OS
Molecular Weight (g/mol) 132.221
MDL Number MFCD09966150
InChI Key LMVCLDAMMNHILO-UHFFFAOYSA-N
Synonym tetrahydro-2h-thiopyran-4-yl methanol, tetrahydrothiopyran-4-methanol, tetrahydrothiopyran-4-ylmethanol, thian-4-yl methanol, 2h-thiopyran-4-methanol, tetrahydro, 2h-thiopyran-4-methanol,tetrahydro, 4-hydroxymethyl tetrahydro-2h-thiopyran, acmc-20dmhk, thian-4-ylmethan-1-ol, 4-hydroxymethyl thiane
PubChem CID 20143654
IUPAC Name thian-4-ylmethanol
SMILES C1CSCCC1CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 132.23
Percent Purity 97%
Chemical Name or Material Tetrahydrothiopyran-4-ylmethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.