missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraisopropyl methylenediphosphonate, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Tetraisopropyl methylenediphosphonate is used in the preparation of AZT 5’-Triphosphate compounds which display inhibitory effects on HIV-1 reverse transcrpitase. It is also used in the preparation of E2-bisphosphonate conjugates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetraisopropyl methylenediphosphonate is used in the preparation of AZT 5’-Triphosphate compounds which display inhibitory effects on HIV-1 reverse transcrpitase. It is also used in the preparation of E2-bisphosphonate conjugates.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Tetraisopropyl methylenediphosphonate is used in the preparation of AZT 5’-Triphosphate compounds which display inhibitory effects on HIV-1 reverse transcrpitase. It is also used in the preparation of E2-bisphosphonate conjugates.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.08 |
Boiling Point | 122°C to 124°C (1.5 mmHg) |
Flash Point | 160°C (320°F) |
Refractive Index | 1.434 |
Quantity | 10 g |
Beilstein | 1080180 |
Solubility Information | Insoluble in water. |
Formula Weight | 344.33 |
Percent Purity | 98% |
Chemical Name or Material | Tetraisopropyl methylenediphosphonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'