missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(1-pyrazolyl)borate, potassium salt, 95%, Thermo Scientific Chemicals

Product Code. 10628324
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14782-58-2
Molecular Formula C12H12BKN8
Molecular Weight (g/mol) 318.2
MDL Number MFCD00800557
InChI Key RKBGLZQVHBLUBM-UHFFFAOYSA-N
Synonym tetrakis 1-pyrazolyl borate, potassium tetrakis pyrazol-1-yloxy boranuide
PubChem CID 92043288
IUPAC Name potassium;tetra(pyrazol-1-yloxy)boranuide
SMILES [B-](ON1C=CC=N1)(ON2C=CC=N2)(ON3C=CC=N3)ON4C=CC=N4.[K+]

Specifications

Melting Point 253°C to 254°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in alcohols,dmf,thf
Formula Weight 318.2
Percent Purity 95%
Physical Form Fine Crystalline Powder
Chemical Name or Material Tetrakis(1-pyrazolyl)borate, potassium salt, 95%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'