missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV), 99.9%, Thermo Scientific™
£34.50 - £125.00
Chemical Identifiers
CAS | 18865-74-2 |
---|---|
Molecular Formula | C44H80O8Zr |
Molecular Weight (g/mol) | 828.34 |
MDL Number | MFCD00145380,MFCD00145380 |
InChI Key | ANBXKEDJQLYCLX-UHFFFAOYSA-N |
Synonym | tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium, zirconium 2,2,6,6-tetramethyl-3,5-heptanedionate, tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium iv, zr tmhd 4, zirconium tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionate, zirconium tetrakis dipivaloylmethanate, zirconium iv 2,2,6,6-tetramethyl-3,5-heptanedionate, 4z-2,2,6,6-tetramethyl-5-tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy zirconio oxy hept-4-en-3-one |
PubChem CID | 50919870 |
IUPAC Name | 5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;zirconium |
SMILES | [Zr].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
18865-74-2 | |
828.34 | |
ANBXKEDJQLYCLX-UHFFFAOYSA-N | |
50919870 | |
[Zr].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C |
C44H80O8Zr | |
MFCD00145380,MFCD00145380 | |
tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium, zirconium 2,2,6,6-tetramethyl-3,5-heptanedionate, tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium iv, zr tmhd 4, zirconium tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionate, zirconium tetrakis dipivaloylmethanate, zirconium iv 2,2,6,6-tetramethyl-3,5-heptanedionate, 4z-2,2,6,6-tetramethyl-5-tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy zirconio oxy hept-4-en-3-one | |
5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;zirconium |
Specifications
18865-74-2 | |
180°C (0.1 mmHg)(sublimation) | |
MFCD00145380,MFCD00145380 | |
ANBXKEDJQLYCLX-UHFFFAOYSA-N | |
5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one;zirconium | |
50919870 | |
99.9% |
318°C to 320°C | |
C44H80O8Zr | |
tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium, zirconium 2,2,6,6-tetramethyl-3,5-heptanedionate, tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionato zirconium iv, zr tmhd 4, zirconium tetrakis 2,2,6,6-tetramethyl-3,5-heptanedionate, zirconium tetrakis dipivaloylmethanate, zirconium iv 2,2,6,6-tetramethyl-3,5-heptanedionate, 4z-2,2,6,6-tetramethyl-5-tris 3z-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy zirconio oxy hept-4-en-3-one | |
[Zr].CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C.CC(C)(C)C(O)=CC(=O)C(C)(C)C | |
828.34 | |
824.3 | |
Tetrakis(2,2,6,6-tetramethyl-3,5-heptanedionato)zirconium(IV) |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures