missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrakis(acetonitrile)palladium(II) tetrafluoroborate, Pd 23.5%, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.25 g
1 g
Description
Acts as a precursor for
Synthesis of dendritic SCS-pincer palladium complexes. Palladium complexes of click ligands. Dipalladium catalysts for use in Heck cross-coupling5, Suzuki cross-coupling, and aldehyde olefination.This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 230°C (decomposition) |
Quantity | 0.25 g |
UN Number | UN2811 |
Sensitivity | Air Sensitive |
Solubility Information | Soluble in water |
IUPAC Name | palladium(2+) tetrakis(acetonitrile) bis(tetrafluoroboranuide) |
Formula Weight | 444.22 |
Concentration or Composition (by Analyte or Components) | Pd 23.5% |
Physical Form | Powder |
Chemical Name or Material | Tetrakis(acetonitrile)palladium(II) tetrafluoroborate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'