Learn More
Tetrakis(hydroxymethyl)phosphonium chloride, approx. 75-85% solution in water, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 389441000
60.40 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
for C-C Bond Formation (Olefination)
Specifications
Tetrakis(hydroxymethyl)phosphonium chloride | |
Green-Yellow or Pink | |
115.0°C | |
Authentic | |
C4H12ClO4P | |
100 mL | |
tetrakis hydroxymethyl phosphonium chloride, thpc, pyroset tkc, retardol c, tetramethylolphosphonium chloride, tetrakis hydroxymethyl phosphanium chloride, unii-58wb2xcf8i, proban cc, tetrakis hydroxymethyl phosphochloride, ccris 317 | |
AKXUUJCMWZFYMV-UHFFFAOYSA-M | |
tetrakis(hydroxymethyl)phosphanium;chloride | |
31298 | |
80.0 to 85.0% |
124-64-1, 7732-18-5 | |
1.3400g/mL | |
96°C | |
Glass bottle | |
MFCD00031687 | |
1.34 | |
Solubility in water: soluble. | |
[Cl-].OC[P+](CO)(CO)CO | |
190.56 | |
190.56 | |
Solution |
Safety and Handling
- Tetrakis(hydroxymethyl)phosphonium chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Substances and mixtures corrosive to metals Category 1
- Reproductive toxicity Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- H361d-Suspected of damaging the unborn child.
- H301-Toxic if swallowed.
- H290-May be corrosive to metals.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
RUO – Research Use Only