missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(trimethylsilyloxy)silane, 96%, Thermo Scientific™

Product Code. 10255191 Shop All Thermo Scientific Chemicals Products
Click to view available options
:
Glass bottle; 5g
This item is not returnable. View return policy

Product Code. 10255191

Brand: Thermo Scientific Acros 338590050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 3555-47-3
Density 0.8680g/mL
Boiling Point 103.0°C to 106.0°C (2.0 mmHg)
Flash Point 80°C
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C12H36O4Si5
Linear Formula Si[OSi(CH3)3]4
MDL Number MFCD00051587
Quantity 5g
Specific Gravity 0.868
Synonym tetrakis trimethylsiloxy silane, tetrakis trimethylsilyl orthosilicate, tetrakis trimethylsilyloxy silane, unii-55l9t9a11i, tetrakis trimethylsilyl silicate, trisiloxane, 1,1,1,5,5,5-hexamethyl-3,3-bis trimethylsiloxy, trisiloxane, 1,1,1,5,5,5-hexamethyl-3,3-bis trimethylsilyl oxy, acmc-1cnqp, tetratrimethylsilyl silicate
InChI Key VNRWTCZXQWOWIG-UHFFFAOYSA-N
SMILES C[Si](C)(C)O[Si](O[Si](C)(C)C)(O[Si](C)(C)C)O[Si](C)(C)C
IUPAC Name tetrakis(trimethylsilyl) silicate
Molecular Weight (g/mol) 384.84
PubChem CID 19086
Formula Weight 384.84
Percent Purity 96%
Chemical Name or Material Tetrakis(trimethylsilyloxy)silane, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Tetrakis(trimethylsilyloxy)silane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.