missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrakis(triphenylphosphine)nickel(0), 95%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
5 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 122.0°C to 124.0°C |
Color | Brown to Brown-Rust |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% |
Packaging | Glass bottle |
Linear Formula | [(C6H5)3P]4Ni |
Quantity | 5 g |
Fieser | 06,570; 07,357; 09,450; 12,467; 13,294; 16,317 |
Formula Weight | 1107.89 |
Percent Purity | 95% |
Show More |
Safety and Handling
Product Identifier
- Tetrakis(triphenylphosphine)nickel(0)
- Danger
- Carcinogenicity Category 2
- Flammable solid Category 1
- Respiratory sensitiser Category 1
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- H228-Flammable solid.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'