Learn More
Tetrakis(triphenylphosphine)palladium(0), 99%, Thermo Scientific Chemicals
£83.11 - £2392.91
Chemical Identifiers
CAS | 14221-01-3 |
---|---|
Molecular Formula | C72H60P4Pd |
Molecular Weight (g/mol) | 1155.59 |
MDL Number | MFCD00010012 |
InChI Key | NFHFRUOZVGFOOS-UHFFFAOYSA-N |
Synonym | tetrakis triphenylphosphine palladium, tetrakis triphenylphosphine palladium 0, pd pph3 4, tetrakis triphenylphosphine palladium o, tetra triphenylphosphine palladium, palladium-tetrakis triphenylphosphine, palladium 0 tetrakis triphenylphosphine, palladium, tetrakis triphenylphosphine-, t-4 |
PubChem CID | 11979704 |
IUPAC Name | palladium;triphenylphosphane |
SMILES | [Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10762051
|
Thermo Scientific Acros
202380010 |
1 g | Glass bottle |
£83.11
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10492391
|
Thermo Scientific Acros
202380050 |
5 g | Glass bottle |
£178.89
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10326382
|
Thermo Scientific Acros
202380250 |
25 g | Glass bottle |
£682.40
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11924361
|
Thermo Scientific Acros
202381000 |
100 g | Glass bottle |
£2392.91
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14221-01-3 | |
1155.59 | |
NFHFRUOZVGFOOS-UHFFFAOYSA-N | |
11979704 | |
[Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 |
C72H60P4Pd | |
MFCD00010012 | |
tetrakis triphenylphosphine palladium, tetrakis triphenylphosphine palladium 0, pd pph3 4, tetrakis triphenylphosphine palladium o, tetra triphenylphosphine palladium, palladium-tetrakis triphenylphosphine, palladium 0 tetrakis triphenylphosphine, palladium, tetrakis triphenylphosphine-, t-4 | |
palladium;triphenylphosphane |
Specifications
14221-01-3 | |
Khaki to Yellow | |
Authentic | |
C72H60P4Pd | |
MFCD00010012 | |
15,9367 | |
Solubility in water: insoluble. | |
[Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1 | |
1155.59 | |
1155.58 | |
Fine Powder or Platelets |
103.0°C to 107.0°C | |
98.5% min. | |
99% | |
[(C6H5)3P]4Pd | |
06,571; 07,357; 08,472; 09,451; 10,384; 11,503; 12,468; 13,34; 14,177; 16,98; 17,327 | |
tetrakis triphenylphosphine palladium, tetrakis triphenylphosphine palladium 0, pd pph3 4, tetrakis triphenylphosphine palladium o, tetra triphenylphosphine palladium, palladium-tetrakis triphenylphosphine, palladium 0 tetrakis triphenylphosphine, palladium, tetrakis triphenylphosphine-, t-4 | |
NFHFRUOZVGFOOS-UHFFFAOYSA-N | |
palladium;triphenylphosphane | |
11979704 | |
99% | |
Tetrakis(triphenylphosphine)palladium(0) |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wash face,hands and any exposed skin thoroughly after handling.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 238-086-9
- Tetrakis(triphenylphosphine)palladium(0)
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H302-Harmful if swallowed.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.