missing translation for 'onlineSavingsMsg'
Learn More

Tetramethyl orthocarbonate, 95%, Thermo Scientific Chemicals

Product Code. 15462789
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1850-14-2
Molecular Formula C5H12O4
Molecular Weight (g/mol) 136.147
MDL Number MFCD00008473
InChI Key AHJWSRRHTXRLAQ-UHFFFAOYSA-N
Synonym tetramethyl orthocarbonate, methane, tetramethoxy, orthocarbonic acid tetramethyl ester, tetramethoxy-methane, tetramethylorthocarbonate, acmc-209emf, tetramethyl ortho carbonate, tetramethyl orthocarbonate, purum gc
PubChem CID 74613
IUPAC Name tetramethoxymethane
SMILES COC(OC)(OC)OC

Specifications

Melting Point -5°C
Color Colorless
Density 1.017 g/mL
Boiling Point 114°C
Flash Point 22°C (72°F)
Linear Formula C(OCH3)4
Refractive Index 1.385
Quantity 1 g
UN Number UN3272
Beilstein 1737682
Sensitivity Moisture sensitive
Formula Weight 136.15
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tetramethyl orthocarbonate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'