Learn More
Tetramethylammonium bromide, 98%, Thermo Scientific Chemicals
£12.95 - £528.40
Chemical Identifiers
CAS | 64-20-0 |
---|---|
Molecular Formula | C4H12BrN |
Molecular Weight (g/mol) | 154.05 |
MDL Number | MFCD00011626 |
InChI Key | DDFYFBUWEBINLX-UHFFFAOYSA-M |
Synonym | tetramethylammonium bromide, tetramethylammoniumbromide, tetramethylazanium bromide, methanaminium, n,n,n-trimethyl-, bromide, n,n,n-trimethylmethanaminium bromide, unii-6l9ne9609p, ammonium, tetramethyl-, bromide, methanaminium, n,n,n-trimethyl-, bromide 1:1, c4h12n.br, acmc-209nkm |
PubChem CID | 66137 |
ChEBI | CHEBI:55317 |
SMILES | [Br-].C[N+](C)(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10762231
|
Thermo Scientific Acros
187630050 |
5 g | Glass Bottle | ||||||
10143870
|
Thermo Scientific Acros
187631000 |
100 g | Plastic bottle |
£30.40
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10205530
|
Thermo Scientific Acros
187635000 |
500 g | Plastic bottle |
£119.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13488428
|
Thermo Scientific Acros
187630025 |
2.5 kg | Plastic bottle | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystChemical Identifiers
64-20-0 | |
154.05 | |
DDFYFBUWEBINLX-UHFFFAOYSA-M | |
66137 | |
[Br-].C[N+](C)(C)C |
C4H12BrN | |
MFCD00011626 | |
tetramethylammonium bromide, tetramethylammoniumbromide, tetramethylazanium bromide, methanaminium, n,n,n-trimethyl-, bromide, n,n,n-trimethylmethanaminium bromide, unii-6l9ne9609p, ammonium, tetramethyl-, bromide, methanaminium, n,n,n-trimethyl-, bromide 1:1, c4h12n.br, acmc-209nkm | |
CHEBI:55317 |
Specifications
64-20-0 | |
Authentic | |
C4H12BrN | |
MFCD00011626 | |
DDFYFBUWEBINLX-UHFFFAOYSA-M | |
tetramethylazanium bromide | |
66137 | |
154.05 | |
Crystalline Powder |
Gray | |
98% | |
(CH3)4NBr | |
tetramethylammonium bromide, tetramethylammoniumbromide, tetramethylazanium bromide, methanaminium, n,n,n-trimethyl-, bromide, n,n,n-trimethylmethanaminium bromide, unii-6l9ne9609p, ammonium, tetramethyl-, bromide, methanaminium, n,n,n-trimethyl-, bromide 1:1, c4h12n.br, acmc-209nkm | |
[Br-].C[N+](C)(C)C | |
154.05 | |
CHEBI:55317 | |
≥97.5% | |
Tetramethylammonium bromide, 98% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 200-581-2
missing translation for 'rtecsNumber' : BS7600000
missing translation for 'tsca' : TSCA
RUO – Research Use Only