missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylammonium chloride, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
Specifications
Specifications
Melting Point | 420.0°C |
Color | White to Ivory |
Infrared Spectrum | Authentic |
Assay Percent Range | 98% min. (Argentometry) |
Packaging | Plastic drum |
Linear Formula | (CH3)4NCl |
Quantity | 5 kg |
Solubility Information | Solubility in water: >60% (20°C). Other solubilities: insoluble in ether, benzene, slightly soluble in ethanol |
Formula Weight | 109.6 |
Vapor Pressure | Negligible |
Show More |
Safety and Handling
Product Identifier
- Tetramethylammonium chloride
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity after single exposure Category 1
- H370-Causes damage to organs.
- H315-Causes skin irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H311-Toxic in contact with skin.
- H300-Fatal if swallowed.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction