missing translation for 'onlineSavingsMsg'
Learn More
Tetramethylammonium hydroxide, 25% w/w aq. soln., Electronic Grade, 99.9999% (metals basis), Thermo Scientific Chemicals Product Code.: 11347987

Tetramethylammonium hydroxide, 25% w/w aq. soln., Electronic Grade, 99.9999% (metals basis), Thermo Scientific Chemicals

Product Code. 11347987
250 mL
Click to view available options
Quantity:
250 mL
1 L
This item is not returnable. View return policy

Product Code. 11347987

Brand: Thermo Scientific Alfa Aesar 020932.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tetramethylammonium hydroxide is used to produce tetramethyl-ammonium azide. It is used as an anisotropic etchant of silicon, basic solvent in the development of acidic photoresist in photolithography process, as surfactant in the synthesis of ferrofluid and as a polarographic reagent. It finds application in the production of organic silicon, where it is used for computer silicon wafer surface brightener and cleaning agent. It is also involved in the purification of some metallic elements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-59-2
Molecular Formula C4H13NO
Molecular Weight (g/mol) 91.154
MDL Number MFCD00008280
InChI Key WGTYBPLFGIVFAS-UHFFFAOYSA-M
Synonym tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxideShow MoreShow Less
PubChem CID 60966
IUPAC Name tetramethylazanium;hydroxide
SMILES C[N+](C)(C)C.[OH-]

Specifications

Color Colorless to Yellow
Density 1.016 g/mL
Boiling Point ∼102°C
Linear Formula (CH3)4NOH
Refractive Index 1.384
Quantity 250 mL
UN Number UN1835
Beilstein 3558708
Sensitivity Air Sensitive
Merck Index 14,9224
Solubility Information Miscible with water.
Formula Weight 91.15
Percent Purity 99.9999%
Grade Electronic
Concentration or Composition (by Analyte or Components) 25% w/w aq. soln.
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Tetramethylammonium hydroxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM30L011 Certificate TypeCertificate of Analysis Date03/02/2025 Catalog Number020932.LU,020932.AK,020932.LV,020932.K2
Lot NumberX15K003 Certificate TypeCertificate of Analysis Date25/10/2024 Catalog Number020932.LU,020932.AK,020932.LV,020932.K2
Lot NumberR03K034 Certificate TypeCertificate of Analysis Date08/05/2024 Catalog Number020932.LU,020932.AK,020932.LV,020932.K2
Lot NumberN01K017 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog Number020932.LU,020932.AK,020932.LV,020932.K2
Lot NumberR14G028 Certificate TypeCertificate of Analysis Date21/06/2022 Catalog Number020932.LU,020932.AK,020932.LV,020932.K2
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetramethylammonium hydroxide, 25% w/w aqueous soluion, Electronic Grade
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H300 + H310-Fatal if swallowed or in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H370-Causes damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to in accordance with local/regional/national/international regulations..

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetramethylammonium hydroxide, 25% w/w aq. soln., Electronic Grade, 99.9999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.