missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium hydroxide, 25% in water, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 420520050

Code : NEW

Additional Details : CAS Number : 75-59-2 Weight : 5.00000kg Transport : UN number : 1835 Chem class : 8 Pack group : II


 View more versions of this product

Product Code. 13448568

  • £471.00 / 5kg
Estimated Shipment: 31-07-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Tetramethylammonium hydroxide
75-59-2,7732-18-5
Colorless to Yellow
102.0°C
Plastic bottle
1.3780 to 1.3840 (20°C, 589nm)
MFCD00008280
04,5
1.014
tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
WGTYBPLFGIVFAS-UHFFFAOYSA-M
tetramethylazanium;hydroxide
60966
91.15
Solution
25% in Water
-25.0°C
1.0140g/mL
>95°C
C4H13NO
(CH3)4NOH
5 kg
11,514
15,9371
Solubility in water: soluble.
C[N+](C)(C)C.[OH-]
91.15
3.13 mPa.s (19°C)
23 to 27%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tetramethylammonium hydroxide, 25% in water

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H301-Toxic if swallowed.
  • H310-Fatal in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

Supplemental information
  • MIXTURE LIST-Contains : Tetramethylammoniumhydroxide

SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only