missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylammonium hydroxide, 25 wt% in methanol, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 138355000
54.56 GBP valid until 2024-12-31
Use promo code "21665" to get your promotional price.
View more versions of this product
This item is not returnable.
View return policy
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
Tetramethylammonium hydroxide | |
75-59-2, 67-56-1 | |
-98.0°C | |
(CH3)4NOH | |
0.9000g/mL | |
Solubility in water: soluble. | |
C[N+](C)(C)C.[OH-] | |
91.154 | |
91.15 | |
0.9 | |
6°C |
25 wt. % in Methanol | |
65.0°C | |
C4H13NO | |
MFCD00008280 | |
tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide | |
WGTYBPLFGIVFAS-UHFFFAOYSA-M | |
tetramethylazanium;hydroxide | |
60966 | |
Plastic bottle | |
500 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction