missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylammonium Hydroxide Solution, TraceSELECT™ Ultra, Honeywell™
Click to view available options
Quantity:
250 mL
Specifications
Specifications
CAS | 75-59-2 |
Density | 1.020g/mL at 20°C |
Molecular Formula | C4H13NO |
Linear Formula | (CH3)4N(OH) |
MDL Number | MFCD00008280 |
Quantity | 250 mL |
UN Number | 1835 |
Beilstein | 3558708 |
Synonym | tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide |
InChI Key | WGTYBPLFGIVFAS-UHFFFAOYSA-M |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'