Learn More
Tetramethylcyclobutane-1,3-dione, 99%, Thermo Scientific Chemicals
£44.10 - £598.00
Chemical Identifiers
CAS | 933-52-8 |
---|---|
Molecular Formula | C8H12O2 |
Molecular Weight (g/mol) | 140.182 |
MDL Number | MFCD00001331 |
InChI Key | RGCDVHNITQEYPO-UHFFFAOYSA-N |
Synonym | tetramethyl-1,3-cyclobutanedione, tetramethylcyclobutane-1,3-dione, 1,3-cyclobutanedione, 2,2,4,4-tetramethyl, tetramethylcyclobuta-1,3-dione, 2,2,4,4-tetramethyl-1,3-cyclobutanedione, unii-rt4aq22ks4, 1,1,3,3-tetramethylcyclobutanedione, 2,2,4,4-tetramethylcyclobutanedione, dimethyl ketene dimer, rt4aq22ks4 |
PubChem CID | 13617 |
IUPAC Name | 2,2,4,4-tetramethylcyclobutane-1,3-dione |
SMILES | CC1(C(=O)C(C1=O)(C)C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11477757
|
Thermo Scientific Alfa Aesar
A16297.09 |
10 g |
£44.10
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11487757
|
Thermo Scientific Alfa Aesar
A16297.18 |
50 g |
£151.00
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11497757
|
Thermo Scientific Alfa Aesar
A16297.30 |
250 g |
£598.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tetramethyl-1,3-cyclobutanedione is used as a chemical for life science research. It is also used to make other chemical, as well as an chemical and organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetramethyl-1,3-cyclobutanedione is used as a chemical for life science research. It is also used to make other chemical, as well as an chemical and organic intermediate.
Solubility
It is soluble in chloroform.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents.
Chemical Identifiers
933-52-8 | |
140.182 | |
RGCDVHNITQEYPO-UHFFFAOYSA-N | |
13617 | |
CC1(C(=O)C(C1=O)(C)C)C |
C8H12O2 | |
MFCD00001331 | |
tetramethyl-1,3-cyclobutanedione, tetramethylcyclobutane-1,3-dione, 1,3-cyclobutanedione, 2,2,4,4-tetramethyl, tetramethylcyclobuta-1,3-dione, 2,2,4,4-tetramethyl-1,3-cyclobutanedione, unii-rt4aq22ks4, 1,1,3,3-tetramethylcyclobutanedione, 2,2,4,4-tetramethylcyclobutanedione, dimethyl ketene dimer, rt4aq22ks4 | |
2,2,4,4-tetramethylcyclobutane-1,3-dione |
Specifications
933-52-8 | |
120°C to 122°C (150 mmHg) | |
MFCD00001331 | |
It is soluble in chloroform. | |
CC1(C(=O)C(C1=O)(C)C)C | |
140.182 | |
140.18 | |
Tetramethylcyclobutane-1,3-dione |
112°C to 115°C | |
C8H12O2 | |
tetramethyl-1,3-cyclobutanedione, tetramethylcyclobutane-1,3-dione, 1,3-cyclobutanedione, 2,2,4,4-tetramethyl, tetramethylcyclobuta-1,3-dione, 2,2,4,4-tetramethyl-1,3-cyclobutanedione, unii-rt4aq22ks4, 1,1,3,3-tetramethylcyclobutanedione, 2,2,4,4-tetramethylcyclobutanedione, dimethyl ketene dimer, rt4aq22ks4 | |
RGCDVHNITQEYPO-UHFFFAOYSA-N | |
2,2,4,4-tetramethylcyclobutane-1,3-dione | |
13617 | |
99% |
Safety and Handling
missing translation for 'einecsNumber' : 213-269-6
missing translation for 'rtecsNumber' : GU1775000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only