missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylphosphonium bromide, 97%, Thermo Scientific Chemicals

Product Code. 11409026
Click to view available options
Quantity:
1 g
5 g
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

It is used as a pharmaceutical intermediate and in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate and in chemical research.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 4519-28-2
Molecular Formula C4H12BrP
Molecular Weight (g/mol) 171.02
MDL Number MFCD00011802
InChI Key ZTXFOCMYRCGSMU-UHFFFAOYSA-M
Synonym tetramethylphosphonium bromide, tetramethylphosphanium bromide, acmc-1ai8x, ch3 4pbr, tetramethyl phosphonium bromide, phosphonium, tetramethyl-, bromide
PubChem CID 357594
IUPAC Name tetramethylphosphanium;bromide
SMILES [Br-].C[P+](C)(C)C

Spécification

Quantity 1 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 171.03
Percent Purity 98%
Chemical Name or Material Tetramethylphosphonium bromide
compliance-icons
Identifiant produit
  • Tetramethylphosphonium bromide
Mention d'avertissement
  • Warning
Catégorie de danger
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Mention de danger
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Conseils de prudence
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Informations supplémentaires
  • MIXTURE LIST-Contains : Tetramethylphosphoni

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'