missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylsilane, 99.9%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Tetramethylsilane is used as a building block in organometallic chemistry. It acts as a by-product in the production of methyl chlorosilanes. Also, it serves as a precursor to silicon dioxide or silicon carbide. It is used as internal reference standard for the calibration of chemical sift for 1, 13 and 29 NMR spectroscopy. In addition, it is used as an aviation fuel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetramethylsilane is used as a building block in organometallic chemistry. It acts as a by-product in the production of methyl chlorosilanes. Also, it serves as a precursor to silicon dioxide or silicon carbide. It is used as internal reference standard for the calibration of chemical sift for 1, 13 and 29 NMR spectroscopy. In addition, it is used as an aviation fuel.
Solubility
Miscible with water, ethanol, ether and most organic solvents. Immiscible with cold sulfuric acid.
Notes
Store in a cool place. Incompatible with strong acids, strong bases and strong oxidizing agents.
Tetramethylsilane is used as a building block in organometallic chemistry. It acts as a by-product in the production of methyl chlorosilanes. Also, it serves as a precursor to silicon dioxide or silicon carbide. It is used as internal reference standard for the calibration of chemical sift for 1, 13 and 29 NMR spectroscopy. In addition, it is used as an aviation fuel.
Solubility
Miscible with water, ethanol, ether and most organic solvents. Immiscible with cold sulfuric acid.
Notes
Store in a cool place. Incompatible with strong acids, strong bases and strong oxidizing agents.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 26°C to 27°C |
Melting Point | -100°C |
Quantity | 25 g |
UN Number | UN2749 |
Beilstein | 1696908 |
Flash Point | −27°C (−16°F) |
Merck Index | 14,9229 |
Solubility Information | Miscible with water,ethanol,ether and most organic solvents. Immiscible with cold sulfuric acid. |
Formula Weight | 88.23 |
Percent Purity | 99.9% |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS09_Aquatic_Toxicity.png)
Signal Word
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 1
- H224-Extremely flammable liquid and vapour.
- H411-Toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'