missing translation for 'onlineSavingsMsg'
Learn More

Tetramethyltin, 98%, Thermo Scientific Chemicals

£20.90 - £252.26

Chemical Identifiers

CAS 594-27-4
Molecular Formula C4H12Sn
Molecular Weight (g/mol) 178.85
MDL Number MFCD00008278
InChI Key VXKWYPOMXBVZSJ-UHFFFAOYSA-N
Synonym tetramethyltin, stannane, tetramethyl, tetramethyl tin, tin, tetramethyl, tetramethylcin, snme4, ch3 4sn, tetramethylcin czech, tin tetramethyl, unii-8v4xu9dpbk
PubChem CID 11661
ChEBI CHEBI:30420
IUPAC Name tetramethylstannane
SMILES C[Sn](C)(C)C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11354839
View Documents Promotion Details
Thermo Scientific Alfa Aesar
071145.04
2 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11344839
View Documents Promotion Details
Thermo Scientific Alfa Aesar
071145.09
10 g
£69.95
10g
Please to purchase this item. Need a web account? Register with us today!
11334839
View Documents Promotion Details
Thermo Scientific Alfa Aesar
071145.18
50 g
£252.26
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

594-27-4
178.85
VXKWYPOMXBVZSJ-UHFFFAOYSA-N
11661
tetramethylstannane
C4H12Sn
MFCD00008278
tetramethyltin, stannane, tetramethyl, tetramethyl tin, tin, tetramethyl, tetramethylcin, snme4, ch3 4sn, tetramethylcin czech, tin tetramethyl, unii-8v4xu9dpbk
CHEBI:30420
C[Sn](C)(C)C

Specifications

594-27-4
1.291g/mL
−12°C (9°F)
C4H12Sn
MFCD00008278
tetramethyltin, stannane, tetramethyl, tetramethyl tin, tin, tetramethyl, tetramethylcin, snme4, ch3 4sn, tetramethylcin czech, tin tetramethyl, unii-8v4xu9dpbk
VXKWYPOMXBVZSJ-UHFFFAOYSA-N
tetramethylstannane
11661
178.83
Liquid
-54°C
74°C to 75°C
Unpleasant
1.4410
UN3384
Insoluble in water; Soluble in organic solvents
C[Sn](C)(C)C
178.85
CHEBI:30420
98%
Tetramethyltin
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H300-H310-H330
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S.

missing translation for 'rtecsNumber' : WH8630000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only