Learn More
Tetraphenylboron sodium, ACS reagent, Thermo Scientific Chemicals
£81.61 - £963.80
Chemical Identifiers
CAS | 143-66-8 |
---|---|
Molecular Formula | C24H20BNa |
Molecular Weight (g/mol) | 342.22 |
MDL Number | MFCD00011494 |
InChI Key | HFSRCEJMTLMDLI-UHFFFAOYSA-N |
Synonym | sodium tetraphenylborate, tetraphenylboron sodium, sodium tetraphenylboron, kalignost, sodium tetraphenylborate 1-, kalibor, dotite kalibor, kariporu k, borate 1-, tetraphenyl-, sodium, sodium tetraphenylboride |
PubChem CID | 2723787 |
IUPAC Name | sodium tetraphenylboranuide |
SMILES | [Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10351171
|
Thermo Scientific Acros
424450250 |
25 g | Glass bottle |
£81.61
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10311221
|
Thermo Scientific Acros
424451000 |
100 g | Glass bottle |
£223.11
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11905881
|
Thermo Scientific Acros
424455000 |
500 g | Glass bottle |
£963.80
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
143-66-8 | |
342.22 | |
HFSRCEJMTLMDLI-UHFFFAOYSA-N | |
2723787 | |
[Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C24H20BNa | |
MFCD00011494 | |
sodium tetraphenylborate, tetraphenylboron sodium, sodium tetraphenylboron, kalignost, sodium tetraphenylborate 1-, kalibor, dotite kalibor, kariporu k, borate 1-, tetraphenyl-, sodium, sodium tetraphenylboride | |
sodium tetraphenylboranuide |
Specifications
>300.0°C | |
Colorless to White | |
C24H20BNa | |
MFCD00011494 | |
sodium tetraphenylborate, tetraphenylboron sodium, sodium tetraphenylboron, kalignost, sodium tetraphenylborate 1-, kalibor, dotite kalibor, kariporu k, borate 1-, tetraphenyl-, sodium, sodium tetraphenylboride | |
HFSRCEJMTLMDLI-UHFFFAOYSA-N | |
sodium tetraphenylboranuide | |
2723787 | |
ACS Reagent | |
0.5 % max. (105°C) |
143-66-8 | |
Crystals or Granular Powder | |
(C6H5)4BNa | |
15,8818 | |
Solubility in water: 50g/L (20°C). Other solubilities: freely soluble in acetone, slightly soluble in ether and chloroform, practically insoluble in petroleum ether | |
[Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
342.22 | |
342.21 | |
Authentic | |
Tetraphenylboron sodium |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective cloth
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 205-605-5
- Tetraphenylboron sodium
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only