missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraphenylcyclopentadienone, 99%, Thermo Scientific Chemicals
£26.03 - £213.06
Chemical Identifiers
CAS | 479-33-4 |
---|---|
Molecular Formula | C29H20O |
Molecular Weight (g/mol) | 384.47 |
MDL Number | MFCD00001407 |
InChI Key | PLGPSDNOLCVGSS-UHFFFAOYSA-N |
Synonym | tetraphenylcyclopentadienone, tetracyclone, cyclone, tetracyclon, 2,3,4,5-tetraphenylcyclopenta-2,4-dienone, 2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenyl, tetraphenyl-2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenylcyclopentadienone, cyclone compound, 2,3,4,5-tetraphenyl-2,4-cyclopentadienone |
PubChem CID | 68068 |
IUPAC Name | 2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-one |
SMILES | C1=CC=C(C=C1)C2=C(C(=O)C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10096103
|
Thermo Scientific Acros
138540050 |
5 g | Glass bottle |
£26.03
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10635382
|
Thermo Scientific Acros
138540250 |
25 g | Glass bottle |
£74.77
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10234710
|
Thermo Scientific Acros
138541000 |
100 g | Glass bottle |
£213.06
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
479-33-4 | |
384.47 | |
PLGPSDNOLCVGSS-UHFFFAOYSA-N | |
68068 | |
C1=CC=C(C=C1)C2=C(C(=O)C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5 |
C29H20O | |
MFCD00001407 | |
tetraphenylcyclopentadienone, tetracyclone, cyclone, tetracyclon, 2,3,4,5-tetraphenylcyclopenta-2,4-dienone, 2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenyl, tetraphenyl-2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenylcyclopentadienone, cyclone compound, 2,3,4,5-tetraphenyl-2,4-cyclopentadienone | |
2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-one |
Specifications
479-33-4 | |
Black | |
98.5% min. (HPLC) | |
MFCD00001407 | |
tetraphenylcyclopentadienone, tetracyclone, cyclone, tetracyclon, 2,3,4,5-tetraphenylcyclopenta-2,4-dienone, 2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenyl, tetraphenyl-2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenylcyclopentadienone, cyclone compound, 2,3,4,5-tetraphenyl-2,4-cyclopentadienone | |
PLGPSDNOLCVGSS-UHFFFAOYSA-N | |
2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-one | |
68068 | |
99% | |
Tetraphenylcyclopentadienone, 99% |
217.0°C to 222.0°C | |
Authentic | |
C29H20O | |
01,1149; 04,490; 06,577 | |
Solubility in water: insoluble. | |
C1=CC=C(C=C1)C2=C(C(=O)C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5 | |
384.47 | |
384.47 | |
Fine Crystalline Powder |
Safety and Handling
LIGHT SENSITIVE
missing translation for 'einecsNumber' : 207-530-3
missing translation for 'tsca' : TSCA
Product Identifier
- Tetraphenylcyclopentadienone
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title