missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenyllead, 97%, Thermo Scientific™

Product Code. 11424883
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 595-89-1
Molecular Formula C24H20Pb
Molecular Weight (g/mol) 515.60
MDL Number MFCD00002999
InChI Key WBJSMHDYLOJVKC-UHFFFAOYSA-N
Synonym tetraphenyllead, tetraphenyl lead, plumbane, tetraphenyl, pbph4, lead, tetraphenyl-,, lead tetraphenyl, acmc-209mel, wbjsmhdylojvkc-uhfffaoysa
PubChem CID 72906
ChEBI CHEBI:30184
IUPAC Name tetraphenylplumbane
SMILES C1=CC=C(C=C1)[Pb](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 227°C to 228°C
Boiling Point 126°C (13 mmHg)
Quantity 25 g
UN Number UN3467
Formula Weight 515.61
Percent Purity 97%
Chemical Name or Material Tetraphenyllead
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'