missing translation for 'onlineSavingsMsg'
Learn More
Tetrapropylammonium chloride, 94%, Thermo Scientific Chemicals Product Code.: 10147143

Tetrapropylammonium chloride, 94%, Thermo Scientific Chemicals

Product Code. 10147143
100 g, Plastic bottle
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10147143

Brand: Thermo Scientific Acros 420711000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5810-42-4
Molecular Formula C12H28ClN
Molecular Weight (g/mol) 221.81
MDL Number MFCD00038729
InChI Key FBEVECUEMUUFKM-UHFFFAOYSA-M
Synonym tetrapropylammonium chloride, tetrapropyl ammonium chloride, tetra-n-propylammonium chloride, 1-propanaminium, n,n,n-tripropyl-, chloride, 1-propanaminium, n,n,n-tripropyl-, chloride 1:1, n,n,n-tripropylpropan-1-aminium chloride, tetrapropylazanium chloride, acmc-209m2k, ksc491c1h, tetra-n-propylammonium chloride 1gShow MoreShow Less
PubChem CID 79880
IUPAC Name tetrapropylazanium;chloride
SMILES CCC[N+](CCC)(CCC)CCC.[Cl-]

Specifications

Melting Point 240.0°C to 242.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 92% min. (Argentometry)
Packaging Plastic bottle
Linear Formula (CH3CH2CH2)4NCl
Quantity 100 g
Formula Weight 221.81
Percent Purity 94%
Physical Form Crystals
Chemical Name or Material Tetrapropylammonium chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0458910 Certificate TypeCertificate of Analysis Date15/02/2024 Catalog Number420710250,420711000
Lot NumberA0455127 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number420710250,420711000
Lot NumberA0448618 Certificate TypeCertificate of Analysis Date13/01/2023 Catalog Number420710250,420711000
Lot NumberA0394857 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number420710250,420711000
Lot NumberA0401801 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number420710250,420711000

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrapropylammonium chloride, 94%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.