missing translation for 'onlineSavingsMsg'
Learn More

Tetrapropylammonium perchlorate, 98%, Thermo Scientific™

Product Code. 10782022
Click to view available options
Quantity:
25 g
100 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Oxidation reagent, Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15780-02-6
Molecular Formula C12H28ClNO4
Molecular Weight (g/mol) 285.8
MDL Number MFCD00041979
InChI Key LJDKWXHIGFOFDB-UHFFFAOYSA-M
Synonym tetrapropylammonium perchlorate, 1-propanaminium, n,n,n-tripropyl-, perchlorate 1:1, 1-propanaminium, n,n,n-tripropyl-, perchlorate, n,n,n-tripropylpropan-1-aminium perchlorate, tetrapropylazanium perchlorate, tetrapropyl ammonium perchlorate, n,n,n-tripropyl-1-propanaminium perchlorate
PubChem CID 85095
IUPAC Name tetrapropylazanium;perchlorate
SMILES CCC[N+](CCC)(CCC)CCC.[O-]Cl(=O)(=O)=O

Specifications

Melting Point 238°C to 240°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (on dry substance)
Packaging Plastic bottle
Linear Formula (CH3CH2CH2)4NClO4
Quantity 25 g
Formula Weight 285.8
Percent Purity 98%
Physical Form Chunks
Chemical Name or Material Tetrapropylammonium perchlorate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetrapropylammonium perchlorate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.