missing translation for 'onlineSavingsMsg'
Learn More

Tetravinyl tin, 95%

Product Code. 10066013 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 10066013

Brand: Thermo Scientific Acros 321080250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1112-56-7 | C8H12Sn | 226.88 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1112-56-7
Molecular Formula C8H12Sn
Molecular Weight (g/mol) 226.88
MDL Number MFCD00008608
InChI Key MZIYQMVHASXABC-UHFFFAOYSA-N
Synonym tetravinyltin, tetravinylstannane, stannane, tetraethenyl, stannane, tetravinyl, tin tetravinyl, sn ch=ch2 4, tetravinyl tin, tetravinyltin iv, tetraethenylstannane, stannane,tetravinyl
PubChem CID 66189
IUPAC Name tetrakis(ethenyl)stannane
SMILES C=C[Sn](C=C)(C=C)C=C

Specifications

Color Colorless to Yellow
Density 1.2400g/mL
Boiling Point 160°C to 163°C
Flash Point 40°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula Sn(CH=CH2)4
Refractive Index 1.5000 to 1.503
Quantity 25 mL
Specific Gravity 1.24
Formula Weight 226.88
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Tetravinyl tin
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.