missing translation for 'onlineSavingsMsg'
Learn More
Tetronic acid, 96%, Thermo Scientific Chemicals Product Code.: 11428016

Tetronic acid, 96%, Thermo Scientific Chemicals

Product Code. 11428016
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11428016

Brand: Thermo Scientific Alfa Aesar A14079.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tetronic acid is used as a precursor of a variety of substituted and ring-fused furans and butenolides. It is an acidic building block for heterocyclic compounds for medicinal applications. It is used for selective O-alkyaltion reactions involving cesium fluoride. It is also used to prepare 3H-furo[3,4-b]quinolin-1-one by reacting with 2-amino-benzaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetronic acid is used as a precursor of a variety of substituted and ring-fused furans and butenolides. It is an acidic building block for heterocyclic compounds for medicinal applications. It is used for selective O-alkyaltion reactions involving cesium fluoride. It is also used to prepare 3H-furo[3,4-b]quinolin-1-one by reacting with 2-amino-benzaldehyde.

Solubility
Soluble in water, dimethyl sulfoxide, acetonitrile and alcohol.

Notes
Incompatible with strong oxidizing agents and strong alkalis (bases).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-57-1
Molecular Formula C4H4O3
Molecular Weight (g/mol) 100.07
MDL Number MFCD00052187
InChI Key JZQBAGOECGRTSA-UHFFFAOYSA-N
SMILES OC1=CC(=O)OC1

Specifications

Melting Point ∽140°C (decomposition)
Assay Percent Range 96%
Beilstein 108442
Synonym tetronic acid, 2,4 3h,5h-furandione, furan-2,4 3h,5h-dione, tetrahydrofuran-2,4-dione, beta-tetronic acid, butanoic acid, 4-hydroxy-3-oxo-, .gamma.-lactone, tetronicacid, 2,4-furandione, tetrahydrofuran-3,5-dione, 3,5-dihydrofuran-2,4-dione
Solubility Information Soluble in water,dimethyl sulfoxide,acetonitrile and alcohol.
IUPAC Name 4-hydroxy-2,5-dihydrofuran-2-one
PubChem CID 521261
Formula Weight 100.07
Percent Purity 96%
Quantity 5 g
Chemical Name or Material Tetronic Acid
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10183530 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14079.14,A14079.06
Lot Number10214786 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14079.14,A14079.06
Lot Number10177667 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14079.14,A14079.06
Lot Number10229354 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14079.14,A14079.06
Lot Number10183531 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14079.14,A14079.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetronic acid, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.