missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Texas Instruments Scientific Calculator, Solar Powered
This Solar Powered Scientific Calculator features a multi-line display, a choice of three solvers, and the ability to perform vectors and matrices.
Brand: Texas Instruments 17311-06
This item is not returnable.
View return policy
Description
- Four-line display.
- One- and two-variable statistics.
- MultiView™ display shows multiple calculations at the same time on screen.
- Select degrees/radians, floating/fix, number format modes.
- Choose from three solvers: numeric equation, polynomial and system of linear equations.
- Display a defined function in a tabular form.
- Determine the numeric derivative and integral for real functions.
- Perform vectors and matrices using a vector and matrix entry window.
Specifications
Scientific Calculator | |
MultiView Display Shows Multiple Calculations at the Same time on Screen | |
TI-36X Pro | |
0.8 in. | |
3.3 in. | |
Solar | |
Slide Case, Instructions |
LCD | |
7 Variables | |
1 Ea. | |
20.32 mm | |
83.82 mm | |
Solar Battery |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Texas Instruments Scientific Calculator, Solar Powered > Solar Powered Scientific Calculator; Model: TI-36X Pro; Memory: 7 Variables
Spot an opportunity for improvement?Share a Content Correction